Learn More
Medchemexpress LLC SR 9186 | 1361414-26-7 | 98.1% | 100 MG
SDP
Catalog No. 5000232956
Encompass_Preferred
SR9186 (ML368) is a selective CYP3A4 inhibitor research compound supplied for in vitro drug metabolism and related studies. It exhibits low-nanomolar potency against CYP3A4-mediated reactions and is provided as solid and solution forms for laboratory use.
  • Selective CYP3A4 inhibitor with reported IC50s of 9 nM, 4 nM, and 38 nM for representative substrates.
  • High purity, typically 98.1%.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • Molecular formula C26H18N6O; molecular weight 430.46 g/mol.
  • Used in drug metabolism studies, and relevant to research on hepatic-stage P. falciparum and breast cancer.
Catalog No. 50-002-32956 Supplier Medchemexpress LLC Supplier No. HY120696100MG
Add to Cart
Add to Cart
Request a Quote
SR9186 (ML368) is a selective CYP3A4 inhibitor research compound supplied for in vitro drug metabolism and related studies. It exhibits low-nanomolar potency against CYP3A4-mediated reactions and is provided as solid and solution forms for laboratory use.
  • Selective CYP3A4 inhibitor with reported IC50s of 9 nM, 4 nM, and 38 nM for representative substrates.
  • High purity, typically 98.1%.
  • Available as solid quantities and as a 10 mM solution in DMSO.
  • Molecular formula C26H18N6O; molecular weight 430.46 g/mol.
  • Used in drug metabolism studies, and relevant to research on hepatic-stage P. falciparum and breast cancer.