Learn More
Apexbio Technology LLC SR 95531 hydrobromide 104104-50-9 50mg
SDP
Catalog No. 5000089017
Encompass_Preferred
SR 95531 hydrobromide (CAS 104104-50-9) also known as Gabazine is a selective competitive antagonist of the GABAA receptor By binding to the GABA recognition site on the receptor it blocks the activation induced by -aminobutyric acid (GABA) Experimental studies report an inhibitory concentration (IC50) of approximately 0 2 M for GABAA receptor-mediated responses Widely utilized in neuroscience research Gabazine is applied to investigate the physiological and pathological roles of GABAA receptor-mediated synaptic transmission neural network modulation and neurotransmitter regulation
Catalog No. 50-000-89017 Supplier Apexbio Technology LLC Supplier No. B666350
SR 95531 hydrobromide (CAS 104104-50-9) also known as Gabazine is a selective competitive antagonist of the GABAA receptor By binding to the GABA recognition site on the receptor it blocks the activation induced by -aminobutyric acid (GABA) Experimental studies report an inhibitory concentration (IC50) of approximately 0 2 M for GABAA receptor-mediated responses Widely utilized in neuroscience research Gabazine is applied to investigate the physiological and pathological roles of GABAA receptor-mediated synaptic transmission neural network modulation and neurotransmitter regulation