Learn More
Cayman Chemical SR 95531 (hydrobromide), Purity: ≥95%, CAS: 104104-50-9, 10 mg
SDP
Catalog No. 502969848
Encompass_Preferred
A derivative of GABA that acts as an antagonist of GABAA receptors (Ki = 74-150 nM); elicits seizures when administered intravenously in mice; effective against GABAA receptor isoforms from mice, rats, and humans
Catalog No. 50-296-9848 Supplier Cayman Chemical Supplier No. 1458510 MG
A derivative of GABA that acts as an antagonist of GABAA receptors (Ki = 74-150 nM); elicits seizures when administered intravenously in mice; effective against GABAA receptor isoforms from mice, rats, and humans