Learn More
Cayman Chemical SR 95531hydrobromIde 25mg
SDP
Catalog No. 502971059
Encompass_Preferred
A derivative of GABA that acts as an antagonist of GABAA receptors (Ki = 74-150 nM); elicits seizures when administered intravenously in mice; effective against GABAA receptor isoforms from mice, rats, and humans
Catalog No. 50-297-1059 Supplier Cayman Chemical Supplier No. 1458525 MG
A derivative of GABA that acts as an antagonist of GABAA receptors (Ki = 74-150 nM); elicits seizures when administered intravenously in mice; effective against GABAA receptor isoforms from mice, rats, and humans