Learn More
Medchemexpress LLC SR-BI/SCARB1 Human | 50UG
SDP
Catalog No. 5000290569
Encompass_Preferred
The SR-BI/SCARB1 protein is a multifunctional receptor that binds a variety of ligands including phospholipids cholesteryl esters lipoproteins phosphatidylserine and apoptotic cells As an HDL receptor it crucially mediates selective cholesterol ether uptake and HDL-dependent cholesterol efflux SR-BI/SCARB1 Protein Human (HEK293 His-Fc) is the recombinant human-derived SR-BI/SCARB1 protein expressed by HEK293 with C-hFc C-His labeled tag
Catalog No. 50-002-90569 Supplier Medchemexpress LLC Supplier No. HYP7665850UG
The SR-BI/SCARB1 protein is a multifunctional receptor that binds a variety of ligands including phospholipids cholesteryl esters lipoproteins phosphatidylserine and apoptotic cells As an HDL receptor it crucially mediates selective cholesterol ether uptake and HDL-dependent cholesterol efflux SR-BI/SCARB1 Protein Human (HEK293 His-Fc) is the recombinant human-derived SR-BI/SCARB1 protein expressed by HEK293 with C-hFc C-His labeled tag