Learn More
Eichrom Industries Sr Resin, 50-100 μm particle size, 25 g bottle, Extraction Chromatography, 1.0M 4,4′(5′)-di-t-butylcyclohexano 18-crown-6 in 1-octanol
SDP
Catalog No. NC1484883
Encompass
Sr Resin, 50-100 μm particle size, 25 g bottle, Extraction Chromatography, 1.0M 4,4′(5′)-di-t-butylcyclohexano 18-crown-6 in 1-octanol
Catalog No. NC1484883 Supplier Eichrom Industries Supplier No. SRB25S
Sr Resin, 50-100 μm particle size, 25 g bottle, Extraction Chromatography, 1.0M 4,4′(5′)-di-t-butylcyclohexano 18-crown-6 in 1-octanol