Learn More
TARGETMOL CHEMICALS INC SR1001 10MG
SDP
Catalog No. 502579802
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR 1001 is an inverse agonist for ROR(alpha)/ROR(gamma) (Kis 172/111 nM). purity: 99%
Catalog No. 50-257-9802 Supplier TARGETMOL CHEMICALS INC Supplier No. T516310MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR 1001 is an inverse agonist for ROR(alpha)/ROR(gamma) (Kis 172/111 nM). purity: 99%