Learn More
Medchemexpress LLC HY-13421 5mg Medchemexpress, SR1001 CAS:1335106-03-0 Purity:>98%
SDP
Catalog No. 502081360
Encompass_Preferred
Medchemexpress, HY-13421 5mg SR1001 CAS:1335106-03-0 SR1001 is a selective RORα and RORγt inverse agonist with Kis 172 and 111 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1360 Supplier Medchemexpress LLC Supplier No. HY134215MG
Medchemexpress, HY-13421 5mg SR1001 CAS:1335106-03-0 SR1001 is a selective RORα and RORγt inverse agonist with Kis 172 and 111 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.