Learn More
Medchemexpress LLC SR10067 | 1380548-02-6 | 99.2% | 465.58 | C31H31NO3 | 50 MG
SDP
Catalog No. 5000227846
Encompass_Preferred
SR10067 is a potent, selective, brain-penetrant REV-ERBα/β agonist used in preclinical research on circadian regulation, metabolic and neuropsychiatric disorders. Reported IC50 values are approximately 160 nM (Rev-Erbβ) and 170 nM (Rev-Erbα).
  • Potent REV-ERB agonist with low-nanomolar activity.
  • Brain-penetrant small molecule suitable for in vivo studies.
  • High purity (≈99.2%) suitable for research applications.
  • Molecular weight 465.58 and formula C31H31NO3.
  • Solid, white to off-white appearance; storage guidelines provided by supplier.
Catalog No. 50-002-27846 Supplier Medchemexpress LLC Supplier No. HY11751650MG
SR10067 is a potent, selective, brain-penetrant REV-ERBα/β agonist used in preclinical research on circadian regulation, metabolic and neuropsychiatric disorders. Reported IC50 values are approximately 160 nM (Rev-Erbβ) and 170 nM (Rev-Erbα).
  • Potent REV-ERB agonist with low-nanomolar activity.
  • Brain-penetrant small molecule suitable for in vivo studies.
  • High purity (≈99.2%) suitable for research applications.
  • Molecular weight 465.58 and formula C31H31NO3.
  • Solid, white to off-white appearance; storage guidelines provided by supplier.