Learn More
Medchemexpress LLC SR10067 | 1380548-02-6 | 99.2% | 465.58 g·mol⁻1 | C31H31NO3 | 25 MG
SDP
Catalog No. 5000226510
Encompass_Preferred
SR10067 is a potent, selective, brain-penetrant REV-ERB agonist used as a research tool to study circadian biology, metabolic regulation, and neuropsychiatric disorders. It has reported IC50 values of approximately 160-170 nM and a molecular weight of 465.58 g·mol⁻1.
  • Potent, selective REV-ERB agonist with low-nanomolar activity.
  • Brain-penetrant small molecule suitable for in vivo studies.
  • High chemical purity (≈99.2% by supplier analysis).
  • Molecular formula C31H31NO3 and molecular weight 465.58 g·mol⁻1.
  • Stable as powder at -20°C for long-term storage.
  • Suitable for receptor assays, cell-based studies, and in vivo pharmacology.
Catalog No. 50-002-26510 Supplier Medchemexpress LLC Supplier No. HY11751625MG
SR10067 is a potent, selective, brain-penetrant REV-ERB agonist used as a research tool to study circadian biology, metabolic regulation, and neuropsychiatric disorders. It has reported IC50 values of approximately 160-170 nM and a molecular weight of 465.58 g·mol⁻1.
  • Potent, selective REV-ERB agonist with low-nanomolar activity.
  • Brain-penetrant small molecule suitable for in vivo studies.
  • High chemical purity (≈99.2% by supplier analysis).
  • Molecular formula C31H31NO3 and molecular weight 465.58 g·mol⁻1.
  • Stable as powder at -20°C for long-term storage.
  • Suitable for receptor assays, cell-based studies, and in vivo pharmacology.