Learn More
TARGETMOL CHEMICALS INC SR10067 5MG
SDP
Catalog No. 502581389
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR10067 is a potent selective and brain penetrant agonist of Rev-Erb(beta)(IC50 = 160 nM) and Rev-Erb(alpha)(IC50 = 170 nM) with anxiolytic activity. purity: 99%
Catalog No. 50-258-1389 Supplier TARGETMOL CHEMICALS INC Supplier No. T129985MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR10067 is a potent selective and brain penetrant agonist of Rev-Erb(beta)(IC50 = 160 nM) and Rev-Erb(alpha)(IC50 = 170 nM) with anxiolytic activity. purity: 99%