Learn More
Medchemexpress LLC HY-117516 5mg Medchemexpress, SR10067 CAS:1380548-02-6 Purity:>98%
SDP
Catalog No. 501932523
Encompass_Preferred
Medchemexpress, HY-117516 5mg SR10067 CAS:1380548-02-6 SR10067 is a potent, selective and brain penetrant Rev-Erbα/β agonist, with IC 50 values are 160 and 170 nM for Rev-Erbβ and Rev-Erbα, respectively. SR10067 has anxiolytic activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2523 Supplier Medchemexpress LLC Supplier No. HY1175165MG
Medchemexpress, HY-117516 5mg SR10067 CAS:1380548-02-6 SR10067 is a potent, selective and brain penetrant Rev-Erbα/β agonist, with IC 50 values are 160 and 170 nM for Rev-Erbβ and Rev-Erbα, respectively. SR10067 has anxiolytic activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.