Learn More
Medchemexpress LLC HY-14422 5mg Medchemexpress, SR1078 CAS:1246525-60-9 Purity:>98%
SDP
Catalog No. 502030533
Encompass_Preferred
Medchemexpress, HY-14422 5mg SR1078 CAS:1246525-60-9 SR1078 is a selective agonist of retinoic acid receptor-related orphan receptor α/γ (RORα/RORγ). SR1078 directly binds to the ligand binding domain of RORα and RORγ and increases the transcriptional activity of these receptors, leading to stimulation of RORα/γ target gene transcription[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0533 Supplier Medchemexpress LLC Supplier No. HY144225MG
Medchemexpress, HY-14422 5mg SR1078 CAS:1246525-60-9 SR1078 is a selective agonist of retinoic acid receptor-related orphan receptor α/γ (RORα/RORγ). SR1078 directly binds to the ligand binding domain of RORα and RORγ and increases the transcriptional activity of these receptors, leading to stimulation of RORα/γ target gene transcription[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.