Learn More
Medchemexpress LLC HY-100793 5mg Medchemexpress, SR12813 CAS:126411-39-0 Purity:>98%
SDP
Catalog No. 501931952
Encompass_Preferred
Medchemexpress, HY-100793 5mg SR12813 CAS:126411-39-0 SR12813 is an inhibitor of 3-hydroxy-3-methylglutaryl-coenzyme A (HMG-CoA) reductase , with an IC 50 value of 0.85 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1952 Supplier Medchemexpress LLC Supplier No. HY1007935MG
Medchemexpress, HY-100793 5mg SR12813 CAS:126411-39-0 SR12813 is an inhibitor of 3-hydroxy-3-methylglutaryl-coenzyme A (HMG-CoA) reductase , with an IC 50 value of 0.85 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.