Learn More
Medchemexpress LLC HY-13439 5mg Medchemexpress, SR144528 CAS:192703-06-3 Purity:>98%
SDP
Catalog No. 502081365
Encompass_Preferred
Medchemexpress, HY-13439 5mg SR144528 CAS:192703-06-3 SR144528 is a potent and selective CB2 receptor antagonist with a Ki of 0.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1365 Supplier Medchemexpress LLC Supplier No. HY134395MG
Medchemexpress, HY-13439 5mg SR144528 CAS:192703-06-3 SR144528 is a potent and selective CB2 receptor antagonist with a Ki of 0.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.