Learn More
Medchemexpress LLC SR15006 | 2505001-54-5 | 100.0% | C16H20ClN3O4S | 10MG
SDP
Catalog No. 5000258212
Encompass_Preferred
SR15006 is a potent small-molecule inhibitor of Krüppel-like factor 5 (KLF5) with reported cellular activity (IC50 = 41.6 nM in DLD-1/pGL4.18hKLF5p cells). It is supplied as a high-purity solid and is available as a ready-to-use DMSO solution for cell-based research applications; intended for laboratory research use only.
  • Potent cellular inhibition of KLF5 (IC50 = 41.6 nM).
  • Suitable for cell-based assays and pharmacological studies.
  • High reported purity (99.95%).
  • Available in multiple pack sizes including a 10 mg option.
  • Offered as a 10 mM solution in DMSO for convenient reconstitution.
Catalog No. 50-002-58212 Supplier Medchemexpress LLC Supplier No. HY13969110MG
SR15006 is a potent small-molecule inhibitor of Krüppel-like factor 5 (KLF5) with reported cellular activity (IC50 = 41.6 nM in DLD-1/pGL4.18hKLF5p cells). It is supplied as a high-purity solid and is available as a ready-to-use DMSO solution for cell-based research applications; intended for laboratory research use only.
  • Potent cellular inhibition of KLF5 (IC50 = 41.6 nM).
  • Suitable for cell-based assays and pharmacological studies.
  • High reported purity (99.95%).
  • Available in multiple pack sizes including a 10 mg option.
  • Offered as a 10 mM solution in DMSO for convenient reconstitution.