Learn More
Medchemexpress LLC SR15006 | 2505001-54-5 | 99.90% | 385.87 | 100 MG
SDP
Catalog No. 5000350916
Encompass_Preferred
SR15006 is an inhibitor of Krüppel-like factor 5 (KLF5) with an IC50 of 41.6 nM in DLD-1/pGL4.18hKLF5p cells. In vitro studies demonstrated that SR15006 (1 μM or 10 μM) causes a significant reduction in the levels of tested cyclins over 72 hours in DLD-1 CRC cells. This product is for research use only.
  • Inhibits Krüppel-like factor 5 (KLF5)
  • Exhibits IC50 of 41.6 nM in DLD-1/pGL4.18hKLF5p cells
  • Reduces cyclin levels in DLD-1 CRC cells
  • For research use only
Catalog No. 50-003-50916 Supplier Medchemexpress LLC Supplier No. HY139691100MG
SR15006 is an inhibitor of Krüppel-like factor 5 (KLF5) with an IC50 of 41.6 nM in DLD-1/pGL4.18hKLF5p cells. In vitro studies demonstrated that SR15006 (1 μM or 10 μM) causes a significant reduction in the levels of tested cyclins over 72 hours in DLD-1 CRC cells. This product is for research use only.
  • Inhibits Krüppel-like factor 5 (KLF5)
  • Exhibits IC50 of 41.6 nM in DLD-1/pGL4.18hKLF5p cells
  • Reduces cyclin levels in DLD-1 CRC cells
  • For research use only