Learn More
Medchemexpress LLC HY-111454 5mg Medchemexpress, SR17018 CAS:2134602-45-0 Purity:>98%
SDP
Catalog No. 501968873
Encompass_Preferred
Medchemexpress, HY-111454 5mg SR17018 CAS:2134602-45-0 SR17018 is an mu-opioid-receptor (MOR) agonist, binding with GTPγS, with an EC50 of 97 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8873 Supplier Medchemexpress LLC Supplier No. HY1114545MG
Medchemexpress, HY-111454 5mg SR17018 CAS:2134602-45-0 SR17018 is an mu-opioid-receptor (MOR) agonist, binding with GTPγS, with an EC50 of 97 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.