Learn More
Medchemexpress LLC (E)-3-(3,4-dichlorophenyl)-N-[2-(4-methylsulfonylpiperazin-1-yl)-2-oxoethyl]prop-2-enamide | 2505001-62-5 | 99.6% | 420.31 g/mol | C16H19Cl2N3O4S | 5 MG
SDP
Catalog No. 502030449
Encompass_Preferred
SR18662 is a small-molecule inhibitor of Krüppel-like factor 5 (KLF5) intended for research use. It demonstrates potent activity (IC50 ≈ 4.4 nM) and is supplied as a white to off-white solid, accompanied by analytical documentation to support experimental use.
  • Potent KLF5 inhibition (IC50 ≈ 4.4 nM).
  • High purity, supplied as a white to off-white solid.
  • Available in small pack sizes for screening and follow-up studies.
  • Includes analytical documentation such as COA, HNMR, and LCMS.
  • Suitable for in vitro and in vivo research applications.
Catalog No. 50-203-0449 Supplier Medchemexpress LLC Supplier No. HY1365305MG
SR18662 is a small-molecule inhibitor of Krüppel-like factor 5 (KLF5) intended for research use. It demonstrates potent activity (IC50 ≈ 4.4 nM) and is supplied as a white to off-white solid, accompanied by analytical documentation to support experimental use.
  • Potent KLF5 inhibition (IC50 ≈ 4.4 nM).
  • High purity, supplied as a white to off-white solid.
  • Available in small pack sizes for screening and follow-up studies.
  • Includes analytical documentation such as COA, HNMR, and LCMS.
  • Suitable for in vitro and in vivo research applications.