Learn More
Medchemexpress LLC HY-16998 5mg Medchemexpress, SR2211 CAS:1359164-11-6 Purity:>98%
SDP
Catalog No. 501969983
Encompass_Preferred
Medchemexpress, HY-16998 5mg SR2211 CAS:1359164-11-6 SR2211 is a potent, selective synthetic RORγ modulator and functions as an inverse agonist, with a Ki of 105 nM and an IC50 of ~320 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9983 Supplier Medchemexpress LLC Supplier No. HY169985MG
Medchemexpress, HY-16998 5mg SR2211 CAS:1359164-11-6 SR2211 is a potent, selective synthetic RORγ modulator and functions as an inverse agonist, with a Ki of 105 nM and an IC50 of ~320 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.