Learn More
Medchemexpress LLC SR2640 hydrochloride 10mg | 146662-42-2 | 10MG
SDP
Catalog No. 5000241443
Encompass_Preferred
SR2640 (hydrochloride) is a potent and selective competitive leukotriene D4/leukotriene E4 antagonist SR2640 can be used for researching the role of leukotrienes in human asthma[1
Catalog No. 50-002-41443 Supplier Medchemexpress LLC Supplier No. HY10761010MG
SR2640 (hydrochloride) is a potent and selective competitive leukotriene D4/leukotriene E4 antagonist SR2640 can be used for researching the role of leukotrienes in human asthma[1