Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SR33805 | 121345-64-0 | 99.8% | 564.74 g/mol | C32H40N2O5S | 5 MG
SDP

Catalog No. 5000222147
Encompass_Preferred

SR33805 is a potent L-type calcium (Ca2+) channel antagonist used in preclinical research to study cardiac function and calcium-channel pharmacology. It selectively inhibits L-type but not T-type Ca2+ channels and exhibits low-nanomolar activity in cellular assays, with solubility and storage data suitable for laboratory use.

  • Selective L-type calcium channel antagonism, sparing T-type channels.
  • Shows nanomolar potency in depolarized and polarized conditions.
  • High purity appropriate for research applications.
  • Soluble in DMSO at high concentrations for in vitro assays.
  • Offered in small pack sizes convenient for assay testing.
  • Recommended storage conditions provided to maintain stability.

Catalog No. 50-002-22147 Supplier Medchemexpress LLC Supplier No. HY1369095MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SR33805 is a potent L-type calcium (Ca2+) channel antagonist used in preclinical research to study cardiac function and calcium-channel pharmacology. It selectively inhibits L-type but not T-type Ca2+ channels and exhibits low-nanomolar activity in cellular assays, with solubility and storage data suitable for laboratory use.

  • Selective L-type calcium channel antagonism, sparing T-type channels.
  • Shows nanomolar potency in depolarized and polarized conditions.
  • High purity appropriate for research applications.
  • Soluble in DMSO at high concentrations for in vitro assays.
  • Offered in small pack sizes convenient for assay testing.
  • Recommended storage conditions provided to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.