Learn More
Medchemexpress LLC SR33805 | 121345-64-0 | MFCD24873169 | 99.8% | C32H40N2O5S | 10MG
SDP
Catalog No. 5000261278
Encompass_Preferred
SR33805 is a research-grade L-type calcium channel antagonist (CAS 121345-64-0) used in cardiovascular and ion-channel studies. It selectively blocks L-type but not T-type Ca2+ channels and shows potent activity (EC50 4.1 nM depolarized, 33 nM polarized). Supplied as a high-purity compound for laboratory research.
  • Selective L-type calcium channel antagonist suitable for electrophysiology and pharmacology studies.
  • Potent activity with low-nanomolar EC50s for sensitive assays.
  • High purity supports reproducible experimental results.
  • Soluble in DMSO for convenient compound preparation.
  • Available in small pack sizes appropriate for research use.
Catalog No. 50-002-61278 Supplier Medchemexpress LLC Supplier No. HY13690910MG
SR33805 is a research-grade L-type calcium channel antagonist (CAS 121345-64-0) used in cardiovascular and ion-channel studies. It selectively blocks L-type but not T-type Ca2+ channels and shows potent activity (EC50 4.1 nM depolarized, 33 nM polarized). Supplied as a high-purity compound for laboratory research.
  • Selective L-type calcium channel antagonist suitable for electrophysiology and pharmacology studies.
  • Potent activity with low-nanomolar EC50s for sensitive assays.
  • High purity supports reproducible experimental results.
  • Soluble in DMSO for convenient compound preparation.
  • Available in small pack sizes appropriate for research use.