Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SR59230A hydrochloride | 1135278-41-9 | 99.8% | 361.91 g/mol | C21H28ClNO2 | 1 ML
SDP

Catalog No. 5000247407
Encompass_Preferred

SR59230A hydrochloride is a selective β3-adrenergic receptor antagonist used in research. It penetrates the blood-brain barrier and exhibits nanomolar potency against β-adrenergic receptor subtypes, suitable for in vitro and in vivo pharmacology studies.

  • Selective β3-adrenergic receptor antagonist with reported IC50s of 40 nM (β3), 408 nM (β1), and 648 nM (β2)
  • Blood-brain barrier penetrating compound suitable for CNS studies
  • High purity (≈99.8%) and consistent analytical profile
  • Available as a 10 mM solution in DMSO (1 mL) and as solid packs
  • Soluble in DMSO (250 mg/mL) and moderately soluble in water (2.5 mg/mL)
  • Molecular weight 361.91 g/mol; molecular formula C21H28ClNO2

Catalog No. 50-002-47407 Supplier Medchemexpress LLC Supplier No. HY10320010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SR59230A hydrochloride is a selective β3-adrenergic receptor antagonist used in research. It penetrates the blood-brain barrier and exhibits nanomolar potency against β-adrenergic receptor subtypes, suitable for in vitro and in vivo pharmacology studies.

  • Selective β3-adrenergic receptor antagonist with reported IC50s of 40 nM (β3), 408 nM (β1), and 648 nM (β2)
  • Blood-brain barrier penetrating compound suitable for CNS studies
  • High purity (≈99.8%) and consistent analytical profile
  • Available as a 10 mM solution in DMSO (1 mL) and as solid packs
  • Soluble in DMSO (250 mg/mL) and moderately soluble in water (2.5 mg/mL)
  • Molecular weight 361.91 g/mol; molecular formula C21H28ClNO2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.