Learn More
Medchemexpress LLC SR8278 | 1254944-66-5 | 99.9% | C18H19NO3S2 | 50 MG
SDP
Catalog No. 5000354945
Encompass_Preferred
SR8278 is a REV-ERBα antagonist that inhibits the transcriptional repression activity of REV-ERBα with an EC50 of 0.47 μM. It is utilized to regulate metabolism in organisms, study biological rhythms, and research Duchenne muscular dystrophy and Alzheimer's disease.
  • Inhibits REV-ERBα transcriptional repression activity (EC50 of 0.47 μM)
  • Regulates metabolism and biological rhythm
  • Supports research on Duchenne muscular dystrophy
  • Supports research on Alzheimer's disease
  • Exerts antidepressant and anxiolytic effects
  • Restores circadian rhythm of mood-related behaviors
  • Alters REV-ERBα and NURR1 binding activities
Catalog No. 50-003-54945 Supplier Medchemexpress LLC Supplier No. HY1441550MG
SR8278 is a REV-ERBα antagonist that inhibits the transcriptional repression activity of REV-ERBα with an EC50 of 0.47 μM. It is utilized to regulate metabolism in organisms, study biological rhythms, and research Duchenne muscular dystrophy and Alzheimer's disease.
  • Inhibits REV-ERBα transcriptional repression activity (EC50 of 0.47 μM)
  • Regulates metabolism and biological rhythm
  • Supports research on Duchenne muscular dystrophy
  • Supports research on Alzheimer's disease
  • Exerts antidepressant and anxiolytic effects
  • Restores circadian rhythm of mood-related behaviors
  • Alters REV-ERBα and NURR1 binding activities