Learn More
TARGETMOL CHEMICALS INC SR8278 5MG
SDP
Catalog No. 502576279
Encompass_Preferred
Also available in 1 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR8278 is an REV-ERB(alpha) antagonist(EC50 = 0.47 uM) blocking activation of the receptor by the synthetic agonist GSK 4112purity: 99%
Catalog No. 50-257-6279 Supplier TARGETMOL CHEMICALS INC Supplier No. T78375MG
Also available in 1 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SR8278 is an REV-ERB(alpha) antagonist(EC50 = 0.47 uM) blocking activation of the receptor by the synthetic agonist GSK 4112purity: 99%