Learn More
Medchemexpress LLC SR9011 hydrochloride | 2070014-94-5 | 99.0% | 515.50 | 1 ML
SDP
Catalog No. 5000393510
Encompass_Preferred
SR9011 hydrochloride is a REV-ERBα/β agonist with IC50s of 790 nM and 560 nM for REV-ERBα and REV-ERBβ, respectively. This compound offers several key features and benefits:
  • Acts as a REV-ERBα/β agonist
  • Suppresses the expression of BMAL1 mRNA in HepG2 cells in a REV-ERBα/β-dependent manner
  • Suppresses proliferation of breast cancer cell lines
  • Appears to pause the cell cycle of breast cancer cells prior to M phase
  • Causes an increase in cells in the G0/G1 phase and a decrease of cells in S and G2/M phase
  • Displays reasonable plasma exposure in vivo
  • Suppresses plasminogen activator inhibitor type 1 gene (Serpine1), cholesterol 7α-hydroxylase (Cyp7a1), and sterol response element binding protein (Srepf1) in a dose-dependent manner in mice
  • Administration to mice results in loss of locomotor activity during the subject dark phase
Catalog No. 50-003-93510 Supplier Medchemexpress LLC Supplier No. HY16988A10MM/1ML
Add to Cart
Add to Cart
SR9011 hydrochloride is a REV-ERBα/β agonist with IC50s of 790 nM and 560 nM for REV-ERBα and REV-ERBβ, respectively. This compound offers several key features and benefits:
  • Acts as a REV-ERBα/β agonist
  • Suppresses the expression of BMAL1 mRNA in HepG2 cells in a REV-ERBα/β-dependent manner
  • Suppresses proliferation of breast cancer cell lines
  • Appears to pause the cell cycle of breast cancer cells prior to M phase
  • Causes an increase in cells in the G0/G1 phase and a decrease of cells in S and G2/M phase
  • Displays reasonable plasma exposure in vivo
  • Suppresses plasminogen activator inhibitor type 1 gene (Serpine1), cholesterol 7α-hydroxylase (Cyp7a1), and sterol response element binding protein (Srepf1) in a dose-dependent manner in mice
  • Administration to mice results in loss of locomotor activity during the subject dark phase