Learn More
Medchemexpress LLC HY-16988A 5mg Medchemexpress, SR9011 (hydrochloride) CAS:2070014-94-5 Purity:>98%
SDP
Catalog No. 502081921
Encompass_Preferred
Medchemexpress, HY-16988A 5mg SR9011 (hydrochloride) CAS:2070014-94-5 SR9011 hydrochloride is a REV-ERBα/β agonist with IC50s of 790 nM and 560 nM for REV-ERBα and REV-ERBβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1921 Supplier Medchemexpress LLC Supplier No. HY16988A5MG
Medchemexpress, HY-16988A 5mg SR9011 (hydrochloride) CAS:2070014-94-5 SR9011 hydrochloride is a REV-ERBα/β agonist with IC50s of 790 nM and 560 nM for REV-ERBα and REV-ERBβ, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.