Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sr9186 | 1361414-26-7 | 98.1% | 5 MG
SDP

Catalog No. 5000230145
Encompass_Preferred

SR9186 (ML368) is a selective CYP3A4 inhibitor research compound supplied as a solid for laboratory use. It is intended for in vitro studies of CYP3A4-mediated metabolism and drug-drug interactions.

  • Selective CYP3A4 inhibition with low-nanomolar IC50 values.
  • High purity (≈98%).
  • Molecular weight 430.46 g/mol; formula C26H18N6O.
  • Supplied as a yellow to brown solid for laboratory handling.
  • Stable under recommended storage conditions (powder: -20°C or 4°C).
  • Supplied in small research quantities suitable for assay development.

Catalog No. 50-002-30145 Supplier Medchemexpress LLC Supplier No. HY1206965MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SR9186 (ML368) is a selective CYP3A4 inhibitor research compound supplied as a solid for laboratory use. It is intended for in vitro studies of CYP3A4-mediated metabolism and drug-drug interactions.

  • Selective CYP3A4 inhibition with low-nanomolar IC50 values.
  • High purity (≈98%).
  • Molecular weight 430.46 g/mol; formula C26H18N6O.
  • Supplied as a yellow to brown solid for laboratory handling.
  • Stable under recommended storage conditions (powder: -20°C or 4°C).
  • Supplied in small research quantities suitable for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.