Learn More
Abcam SR95531 (Gabazine) (mM/ml), GABAA antagonist, 10MM_ML
Catalog No. 502705675
Encompass_Preferred
MW 368.23 Da, Purity >98%. Selective, competitive GABAA receptor antagonist. Allosteric inhibitor of channel opening of the GABAA receptor. Displaces [3H]-GABA from rat brain membranes with a Ki of 150 nM. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5675 Supplier Abcam Supplier No. AB14448710MM_ML
MW 368.23 Da, Purity >98%. Selective, competitive GABAA receptor antagonist. Allosteric inhibitor of channel opening of the GABAA receptor. Displaces [3H]-GABA from rat brain membranes with a Ki of 150 nM. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement