Learn More
Aviva Systems Biology Corp Sra1 Antibody - N-terminal region (ARP37345_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37345_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sra1. It was validated on Western Blot. Sra1,steroid receptor RNA activator 1,Sra; Srap; Straa1; Strra1; AA959952; Sra1,steroid receptor RNA activator 1,steroid receptor RNA activator 1,steroid receptor RNA activator proteinThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AA959952, SRA, STRAA1, Strra1, Sra, Srap, Straa1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-617