Learn More
Eurofins Discoverx Src (1-530), active, 250 µg. (14-746M)
Supplier:  Eurofins Discoverx 14746M
Encompass
N-terminal 6xHis-tagged recombinant, human Src amino acids 1-530. The mutant form of Src (Src, 1-530) has been identified in cases of advanced colon cancer. This truncating mutation generates an open active form of Src that has been found to be transforming, tumorigenic and metastasis promoting. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9280