Learn More
Carnabio Usa Inc SRC/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0817310
Encompass
SRC, Full-length, Wild type, Amino Acid 1-536, NP_005408.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0991