Learn More
Aviva Systems Biology Corp SRC Antibody - N-terminal region (ARP32475_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32475_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SRC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SRC. Alternate names: ASV, SRC1, c-SRC, p60-SrcApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54612