Learn More
Medchemexpress LLC Src Human sf9 His 10ug
SDP
Catalog No. 5000180309
Encompass_Preferred
Src is a non-receptor protein tyrosine kinase that centralizes multiple cellular processes triggered by various receptors It controls gene transcription immune response adhesion cell cycle apoptosis migration and transformation Src Protein Human (sf9 His-GST) is the recombinant human-derived Src protein expressed by Sf9 insect cells with N-His N-GST labeled tag
Catalog No. 50-001-80309 Supplier Medchemexpress LLC Supplier No. HYP7342210UG
Src is a non-receptor protein tyrosine kinase that centralizes multiple cellular processes triggered by various receptors It controls gene transcription immune response adhesion cell cycle apoptosis migration and transformation Src Protein Human (sf9 His-GST) is the recombinant human-derived Src protein expressed by Sf9 insect cells with N-His N-GST labeled tag