Learn More
TARGETMOL CHEMICALS INC SRC INHIBITOR 1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T359325MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Src Inhibitor 1 (Src Kinase Inhibitor 1) is a potent and selective dual site Src tyrosine kinase inhibitor. purity: 100%
Catalog No. 50-258-0202