Learn More
Medchemexpress LLC SRC INHIBITOR 1 5MG
SDP
Catalog No. NC3440443
Encompass_Preferred
Src Inhibitor 1 is a potent, ATP-competitive and selective dual site Src tyrosine kinase inhibitor with IC50 values of 44 nM for Src and 88nM for Lck.CAS No. : 179248-59-0
Catalog No. NC3440443 Supplier Medchemexpress LLC Supplier No. HY1010535MG
Src Inhibitor 1 is a potent, ATP-competitive and selective dual site Src tyrosine kinase inhibitor with IC50 values of 44 nM for Src and 88nM for Lck.CAS No. : 179248-59-0