Learn More
Selleck Chemical LLC Src Inhibitor 1-5mg
SDP
Supplier:  Selleck Chemical LLC S65675MG
Encompass_Preferred
Src Inhibitor 1 is a potent competitive inhibitor of both Src and Lck (IC50 = 44 and 88 nM, respectively), as well as Csk and Yes.
Catalog No. 50-238-9651