Learn More
Cayman Chemical Src Kinase Inhibitor I, Purity: ≥98%, CAS: 179248-59-0, 1 mg
SDP
Catalog No. 502969530
Encompass_Preferred
A potent competitive inhibitor of both Src and Lck (IC50 = 44 and 88 nM, respectively), as well as Csk and Yes; also inhibits RIP-2
Catalog No. 50-296-9530 Supplier Cayman Chemical Supplier No. 145921 MG
A potent competitive inhibitor of both Src and Lck (IC50 = 44 and 88 nM, respectively), as well as Csk and Yes; also inhibits RIP-2