Learn More
Cayman Chemical Src KInase InhIbItr I 50mg
SDP
Catalog No. 502971921
Encompass_Preferred
A potent competitive inhibitor of both Src and Lck (IC50 = 44 and 88 nM, respectively), as well as Csk and Yes; also inhibits RIP-2
Catalog No. 50-297-1921 Supplier Cayman Chemical Supplier No. 1459250 MG
Add to Cart
Add to Cart
A potent competitive inhibitor of both Src and Lck (IC50 = 44 and 88 nM, respectively), as well as Csk and Yes; also inhibits RIP-2