Learn More
Enzo Life Sciences Src monoclonal antibody (5A18) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905678100
Encompass_Preferred
Clone: 5A18. Host: Mouse. Immunogen: Recombinant human full length c-Src. UniProt: P12931. Source: Purified from hybridoma tissue culture supernatant. Species Reactivity: Human, Mouse, Rat. Applications: IHC (PS), WB. Recommended dilutions: Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Catalog No. 50-658-432