Learn More
Sino Biological Peptide Substrates: SRC Substrate
Catalog No. 502567810
Encompass_Preferred
The SRC sub peptide sequence (KVEKIGEGTYGVVYK-amide) is based on the p34cdc2 (amino acid 6-20).
Catalog No. 50-256-7810 Supplier Sino Biological Supplier No. S3058
The SRC sub peptide sequence (KVEKIGEGTYGVVYK-amide) is based on the p34cdc2 (amino acid 6-20).