Learn More
Enzo Life Sciences Src substrate (1mg)
SDP
Catalog No. 502011275
Encompass_Preferred
This peptide, KVEKIGEGTYGVVYK, is derived from amino acids 6-20 of cdc2 and is a good substrate for src and other tyrosine kinases. Alternative name: cdc2 (6-20). Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1275 Supplier Enzo Life Sciences Supplier No. BMLP1970001
This peptide, KVEKIGEGTYGVVYK, is derived from amino acids 6-20 of cdc2 and is a good substrate for src and other tyrosine kinases. Alternative name: cdc2 (6-20). Purity: ≥95%. Long Term Storage: -20°C.