Learn More
Enzo Life Sciences Src substrate 2 (1mg)
SDP
Catalog No. 502011318
Encompass_Preferred
This peptide, YIYGSFK, was derived through a random combinatorial peptide library method and is a specific and efficient substrate for the scr family of kinases. The Km of YIYGSFK for p60c-src is 55µM. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1318 Supplier Enzo Life Sciences Supplier No. BMLP2590001
This peptide, YIYGSFK, was derived through a random combinatorial peptide library method and is a specific and efficient substrate for the scr family of kinases. The Km of YIYGSFK for p60c-src is 55µM. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.