Learn More
Eurofins Discoverx Src (T341M), active, 10 µg. (14-748)
Supplier:  Eurofins Discoverx 14748
Encompass
N-terminal 6xHis-tagged recombinant, human full length Src, containing the T341M mutation. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9281