Learn More
Cayman Chemical SrchumnrecombInt 10ug
SDP
Catalog No. 502946406
Encompass_Preferred
Source: Active recombinant human N-terminal GST-tagged Src expressed in insect cells • Amino acids: 1-536 • MW: 86 kDa
Catalog No. 50-294-6406 Supplier Cayman Chemical Supplier No. 3256510 UG
Source: Active recombinant human N-terminal GST-tagged Src expressed in insect cells • Amino acids: 1-536 • MW: 86 kDa