Learn More
Abeomics Sre Reporter Hek293 Cell Line
Catalog No. NC1745347
Encompass_Preferred
The SRE Leeporter Luciferase Reporter cell line is a stably transfected HEK 293 cell line which expresses Renilla luciferase reporter gene under the transcriptional control of the serum response element (SRE). The SRE reporter cell line is designed to monitor MAPK/ERK activity and can be used for studying GPCR-linked MAPK/ERK signaling pathways as well as screening of agonists, antagonists or signaling inhibitors related with the MAPK/ERK signaling pathways. Functional activity of the cell line has been validated by serum stimulation assay. Cell line application: (1) Monitor the GPCR-linked MAPK/ERK signaling pathway, and (2) Screen for activators or inhibitors of the GPCR-linked MAPK/ERK signaling pathway. .
Catalog No. NC1745347 Supplier Abeomics Supplier No. 14132ACL
The SRE Leeporter Luciferase Reporter cell line is a stably transfected HEK 293 cell line which expresses Renilla luciferase reporter gene under the transcriptional control of the serum response element (SRE). The SRE reporter cell line is designed to monitor MAPK/ERK activity and can be used for studying GPCR-linked MAPK/ERK signaling pathways as well as screening of agonists, antagonists or signaling inhibitors related with the MAPK/ERK signaling pathways. Functional activity of the cell line has been validated by serum stimulation assay. Cell line application: (1) Monitor the GPCR-linked MAPK/ERK signaling pathway, and (2) Screen for activators or inhibitors of the GPCR-linked MAPK/ERK signaling pathway. .