Learn More
BIOHIPPO SRE-TAL-Puro LV
SDP
Catalog No. 5000331147
Encompass
SRE-TAL-Puro is a transcription factor reporter lentiviral vector designed to monitor MAPK/ERK in living cells This construct delivers a Firefly Luc reporter cassette together with a Puromycin selection marker for generation of stable cell pools The vector is provided as a ready to use Lentivirus preparation pseudotyped with VSV-G for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 2x10E6 transducing units TU Typical applications include Monitor MAPK/ERK pathway activity The reporter is also activated by serum and growth factors The fluorescent reporter enables convenient readout using flow cytometry fluorescence microscopy etc The system is suitable for use with Human/mouse and can support assay development high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-31147 Supplier BIOHIPPO Supplier No. BHV194000243
SRE-TAL-Puro is a transcription factor reporter lentiviral vector designed to monitor MAPK/ERK in living cells This construct delivers a Firefly Luc reporter cassette together with a Puromycin selection marker for generation of stable cell pools The vector is provided as a ready to use Lentivirus preparation pseudotyped with VSV-G for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 2x10E6 transducing units TU Typical applications include Monitor MAPK/ERK pathway activity The reporter is also activated by serum and growth factors The fluorescent reporter enables convenient readout using flow cytometry fluorescence microscopy etc The system is suitable for use with Human/mouse and can support assay development high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures