Learn More
Aviva Systems Biology Corp SREBF1 Antibody - N-terminal region (ARP37145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SREBF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse SREBF1. Alternate names: ADD1, ADD-1, SREBP1, bHLHd1, SREBP-1, SREBP1c, SREBP-1a, SREBP-1c, D630008H06ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56326