Learn More
Aviva Systems Biology Corp SREBF1 Antibody - N-terminal region (ARP38419_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38419_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SREBF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SREBF1. Alternate names: SREBP1, bHLHd1, SREBP-1cApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56647